ADV465748546 AChemBlock

2,4-Dichloro-5,6,7,8-tetrahydro-pyrido[3,4-d]pyrimidine hydrochloride

  • MDL number MFCD11518996
  • Cas Number : 1187830-76-7
  • Molecular Weight : 240.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-4-dichloro-5-6-7-8-tetrahydro-pyrido-3-4-d-pyrimidine-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9484
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart