ADV465748552 AChemBlock

5-Bromo-2-chloro-4-ethoxypyrimidine

  • MDL number MFCD21789322
  • Cas Number : 1289005-72-6
  • Molecular Weight : 237.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-2-chloro-4-ethoxypyrimidine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9490
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart