ADV465748555 AChemBlock

benzyl tert-butyl ((1S,2R,4S)-4-(dimethylcarbamoyl)cyclohexane-1,2-diyl)dicarbamate

  • MDL number MFCD22190576
  • Cas Number : 365998-35-2
  • Molecular Weight : 419.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/carbamic-acid-n-1s-2r-4s-4-dimethylamino-carbonyl-2-1-1-dimethylethoxy-carbonyl-amino-cyclohexyl-phenylmethyl-ester.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9493
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart