ADV465748565 AChemBlock

trans-tert-Butyl 3-hydroxy-2,2,4,4-(tetramethyl)cyclobutylcarbamate

  • MDL number MFCD20923769
  • Cas Number : 1338812-41-1
  • Molecular Weight : 243.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/trans-tert-butyl-3-hydroxy-2-2-4-4-tetramethyl-cyclobutylcarbamate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9503
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart