ADV465748568 AChemBlock

tert-Butyl (2S)-2-(aminomethyl)-2-propylazetidine-1-carboxylate

  • MDL number MFCD28166305
  • Cas Number : 1630815-51-8
  • Molecular Weight : 228.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2s-2-aminomethyl-2-propylazetidine-1-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9506
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart