ADV465748569 AChemBlock

1H-Pyrazolo[4,3-b]pyridine-6-carboxylic acid

  • MDL number MFCD18250815
  • Cas Number : 1256807-59-6
  • Molecular Weight : 163.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form Pink solid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1h-pyrazolo-4-3-b-pyridine-6-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9507
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart