ADV465748570 AChemBlock

5-Chloro-3-methyl-4-amino-1H-pyrazole

  • MDL number MFCD17015936
  • Cas Number : 1064687-15-5
  • Molecular Weight : 131.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/5-chloro-3-methyl-4-amino-1h-pyrazole.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9508
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart