ADV465748575 AChemBlock

(2S)-1-Boc-2-ethylpyrrolidine-2-methanamine

  • MDL number MFCD27987098
  • Cas Number : 1630815-50-7
  • Molecular Weight : 228.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2s-1-boc-2-ethylpyrrolidine-2-methanamine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9513
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart