ADV465748578 AChemBlock

1-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazolo[3,4-b]pyridine

  • MDL number MFCD16995953
  • Cas Number : 1312312-78-9
  • Molecular Weight : 259.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Physical Form White solid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/1-methyl-1h-pyrazolo-3-4-b-pyridine-5-boronic-acid-pinacol-ester.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9516
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart