ADV465748588 AChemBlock

6-(trifluoromethyl)-2-Azaspiro[3.3]heptan-6-ol hydrochloride

  • MDL number MFCD22369882
  • Cas Number : 1384431-10-0
  • Molecular Weight : 217.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/6-trifluoromethyl-2-azaspiro-3-3-heptan-6-ol-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9528
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart