ADV465748591 AChemBlock

3-bromo-1-(tetrahydro-2H-pyran-2-yl)-1H-pyrazolo[3,4-b]pyridine

  • MDL number MFCD22689751
  • Cas Number : 1416713-33-1
  • Molecular Weight : 282.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1h-pyrazolo-3-4-b-pyridine-3-bromo-1-tetrahydro-2h-pyran-2-yl.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9531
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart