ADV465748597 AChemBlock

5-(2-Bromoethyl)-1,3-difluoro-2-methoxybenzene

  • MDL number MFCD27988081
  • Cas Number : 1630906-97-6
  • Molecular Weight : 251.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/5-2-bromoethyl-1-3-difluoro-2-methoxybenzene.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9537
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart