ADV465748600 AChemBlock

8-(Boc-amino)-3-azabicyclo[3.2.1]octane

  • MDL number MFCD18249870
  • Cas Number : 198210-17-2
  • Molecular Weight : 226.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/8-boc-amino-3-azabicyclo-3-2-1-octane.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9541
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart