ADV465748618 AChemBlock

6-Amino-3-bromo-1-methyl-1H-indazole

  • MDL number MFCD12827698
  • Cas Number : 1203181-56-9
  • Molecular Weight : 226.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/6-amino-3-bromo-1-methyl-1h-indazole.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9559
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart