ADV465748620 AChemBlock

3,3-Difluoro-4-hydroxypiperidine hydrochloride

  • MDL number MFCD19686686
  • Cas Number : 1334416-77-1
  • Molecular Weight : 173.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/3-3-difluoro-4-hydroxypiperidine-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9561
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart