ADV465748629 AChemBlock

5-(3,3-Dimethylcyclobutyl)-1H-pyrazol-3-ylamine

  • MDL number MFCD27988085
  • Cas Number : 1853338-91-6
  • Molecular Weight : 165.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/5-3-3-dimethylcyclobutyl-1h-pyrazol-3-ylamine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9571
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart