ADV465748633 AChemBlock

rel-(3R,4R)-1-(tert-butoxycarbonyl)-4-methylpyrrolidine-3-carboxylic acid

  • MDL number MFCD02179038
  • Cas Number : 1253791-53-5
  • Molecular Weight : 229.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1-3-pyrrolidinedicarboxylic-acid-4-methyl-1-1-1-dimethylethyl-ester-3r-4r-rel.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9575
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart