ADV465748640 AChemBlock

Tetrahydro-1H-pyrrolizine-7a(5H)-acetic acid hydrochloride

  • MDL number MFCD09032985
  • Cas Number : 124655-63-6
  • Molecular Weight : 205.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/tetrahydro-1h-pyrrolizine-7a-5h-acetic-acid-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9582
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart