ADV465748643 AChemBlock

(1R,4R)-2-Oxa-5-azabicyclo[2.2.1]heptane

  • MDL number MFCD18374317
  • Cas Number : 661470-56-0
  • Molecular Weight : 99.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1r-4r-2-oxa-5-azabicyclo-2-2-1-heptane.html

Safety Information

signalword Warning
pcodes P233;P244;P303+361+353
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9586
pictograms GHS02
hstatements H226;H302;H311;H314
Availability details for
Quantity
  • Availability
  • Add to Cart