ADV465748656 AChemBlock

2-{[(5Z)-2-(1,2-diazinan-1-yl)-4-oxo-4,5-dihydro-1,3-thiazol-5-ylidene]methyl}-5-fluorophenyl pyrrolidine-1-carboxylate

  • MDL number MFCD28166314
  • Cas Number : 1181083-81-7
  • Molecular Weight : 404.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/2-5z-2-1-2-diazinan-1-yl-4-oxo-4-5-dihydro-1-3-thiazol-5-ylidene-methyl-5-fluorophenyl-pyrrolidine-1-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9599
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart