ADV465748657 AChemBlock

1,9-Diazaspiro[5.5]undecan-2-one hydrochloride

  • MDL number MFCD09998743
  • Cas Number : 1171417-47-2
  • Molecular Weight : 204.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1-9-diazaspiro-5-5-undecan-2-one-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9600
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart