ADV465748662 AChemBlock

4-Cyano-1-piperidinesulfonylchloride

  • MDL number MFCD23106264
  • Cas Number : 1392803-35-8
  • Molecular Weight : 208.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Physical Form White solid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/4-cyano-1-piperidinesulfonylchloride.html
Availability details for
Quantity
  • Availability
  • Add to Cart