ADV465748663 AChemBlock

5-Chloropyrazine-2-carboxaldehyde

  • MDL number MFCD10697774
  • Cas Number : 88625-24-5
  • Molecular Weight : 142.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/5-chloropyrazine-2-carboxaldehyde.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9606
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart