ADV465748667 AChemBlock

4-Amino-6-bromopyrrolo[1,2-f][1,2,4]triazine

  • MDL number MFCD18427507
  • Cas Number : 937047-06-8
  • Molecular Weight : 213.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/4-amino-6-bromopyrrolo-1-2-f-1-2-4-triazine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9610
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart