ADV465748668 AChemBlock

6-Boc-1,6-diaza-spiro[3.5]nonane hemioxalate

  • MDL number MFCD28166317
  • Cas Number : 1630906-45-4
  • Molecular Weight : 542.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/6-boc-1-6-diaza-spiro-3-5-nonane-hemioxalate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9611
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart