ADV465748669 AChemBlock

3-Boc-6-amino-3-azabicyclo[3.2.0]heptane

  • MDL number MFCD19686113
  • Cas Number : 1250884-66-2
  • Molecular Weight : 212.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/3-boc-6-amino-3-azabicyclo-3-2-0-heptane.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9612
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart