ADV465748690 AChemBlock

(2R)-4-Oxoazetidine-2-carboxylic acid

  • MDL number MFCD16036189
  • Cas Number : 62860-12-2
  • Molecular Weight : 115.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2r-4-oxoazetidine-2-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9634
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart