ADV465748691 AChemBlock

(3S,5R)-3-(Boc-amino)-5-methylpiperidine

  • MDL number MFCD18072962
  • Cas Number : 1203651-07-3
  • Molecular Weight : 214.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/3s-5r-3-boc-amino-5-methylpiperidine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9635
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart