ADV465748710 AChemBlock

(7R)-2-chloro-8-cyclopentyl-7-ethyl-7,8-dihydro-5-methyl-6(5H)-pteridinone

  • MDL number MFCD10698781
  • Cas Number : 755039-55-5
  • Molecular Weight : 294.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/7r-2-chloro-8-cyclopentyl-7-ethyl-7-8-dihydro-5-methyl-6-5h-pteridinone.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9654
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart