ADV465748716 AChemBlock

trans-3-(3-Fluoro-4-methoxyphenoxy)cyclobutan-1-amine hydrochloride

  • MDL number MFCD28166325
  • Cas Number : 1630906-63-6
  • Molecular Weight : 247.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/trans-3-3-fluoro-4-methoxyphenoxy-cyclobutan-1-amine-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9660
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart