ADV465748748 AChemBlock

2-chloro-7H-Pyrrolo[2,3-d]pyrimidine-5-carboxylic acid methyl ester

  • MDL number MFCD20923472
  • Cas Number : 1352396-67-8
  • Molecular Weight : 211.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-chloro-7h-pyrrolo-2-3-d-pyrimidine-5-carboxylic-acid-methyl-ester.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9694
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart