ADV465748750 AChemBlock

tert-Butyl 5-oxo-2-azaspiro[3.4]octane-2-carboxylate

  • MDL number MFCD28155163
  • Cas Number : 1581683-57-9
  • Molecular Weight : 225.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-5-oxo-2-azaspiro-3-4-octane-2-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9696
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart