ADV465748760 AChemBlock

3-Aminomethyl-3-(pyrrolidin-1-yl)oxetane

  • MDL number MFCD22628718
  • Cas Number : 1416323-30-2
  • Molecular Weight : 156.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-aminomethyl-3-pyrrolidin-1-yl-oxetane.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9707
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart