ADV465748768 AChemBlock

2-(3-Hydroxyazetidin-3-yl)piperidine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD19439669
  • Cas Number : 934666-06-5
  • Molecular Weight : 256.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/2-3-hydroxyazetidin-3-yl-piperidine-1-carboxylic-acid-tert-butyl-ester.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9715
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart