ADV465748774 AChemBlock

N1-(4-Chlorophenyl)-6-methyl-N5-[3-(9H-purin-6-yl)-2-pyridinyl]-1,5-isoquinolinediamine dihydrochloride

  • MDL number MFCD18782676
  • Cas Number : 1191385-19-9
  • Molecular Weight : 551.87

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/n1-4-chlorophenyl-6-methyl-n5-3-9h-purin-6-yl-2-pyridinyl-1-5-isoquinolinediamine-dihydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9721
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart