ADV465748776 AChemBlock

tert-Butyl N-(4-oxocyclopent-2-en-1-yl)carbamate

  • MDL number MFCD09954963
  • Cas Number : 657396-97-9
  • Molecular Weight : 197.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-4-oxocyclopent-2-en-1-yl-carbamate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9723
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart