ADV465748779 AChemBlock

3,8-Dibromo-1,5-naphthyridine

  • MDL number MFCD22566247
  • Cas Number : 154015-16-4
  • Molecular Weight : 287.94

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/3-8-dibromo-1-5-naphthyridine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9727
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart