ADV465748784 AChemBlock

4-Chloroimidazo[1,5-f][1,2,4]triazine

  • MDL number MFCD08166405
  • Cas Number : 889945-79-3
  • Molecular Weight : 154.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/4-chloroimidazo-1-5-f-1-2-4-triazine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9732
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart