ADV465748796 AChemBlock

(R)-Tetrahydro-2H-pyran-3-ylamine hydrochloride

  • MDL number MFCD09056863
  • Cas Number : 1315500-31-2
  • Molecular Weight : 137.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/r-tetrahydro-2h-pyran-3-ylamine-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9746
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart