ADV465748814 AChemBlock

trans-(4-Fluoro-2-methoxyphenoxy)cyclobutan-1-amine hydrochloride

  • MDL number MFCD28166340
  • Cas Number : 1630906-51-2
  • Molecular Weight : 247.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/trans-4-fluoro-2-methoxyphenoxy-cyclobutan-1-amine-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9765
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart