ADV465748817 AChemBlock

(R)-2-(Dimethylamino)methylpyrrolidine

  • MDL number MFCD19160627
  • Cas Number : 1166842-56-3
  • Molecular Weight : 128.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/r-2-dimethylamino-methylpyrrolidine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9768
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart