ADV465748825 AChemBlock

2,4-Dichloro-7H-Pyrrolo[2,3-d]pyrimidine-5-carbonitrile

  • MDL number MFCD12828241
  • Cas Number : 1379367-43-7
  • Molecular Weight : 213.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-4-dichloro-7h-pyrrolo-2-3-d-pyrimidine-5-carbonitrile.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9778
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart