ADV465748831 AChemBlock

1H-Pyrazolo[3,4-d]pyrimidine

  • MDL number MFCD06407902
  • Cas Number : 271-80-7
  • Molecular Weight : 120.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/1h-pyrazolo-3-4-d-pyrimidine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9785
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart