ADV465748846 AChemBlock

methyl 1-methyl-6-oxo-1,6-dihydropyridazine-3-carboxylate

  • MDL number MFCD23716634
  • Cas Number : 74173-58-3
  • Molecular Weight : 168.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-pyridazinecarboxylic-acid-1-6-dihydro-1-methyl-6-oxo-methyl-ester.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9800
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart