ADV465748849 AChemBlock

3-Cyano-1H-pyrazolo[3,4-c]pyridine

  • MDL number MFCD13176780
  • Cas Number : 245325-34-2
  • Molecular Weight : 144.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/3-cyano-1h-pyrazolo-3-4-c-pyridine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9803
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart