ADV465748852 AChemBlock

methyl 2-(5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)acetate

  • MDL number MFCD18255618
  • Cas Number : 1428761-14-1
  • Molecular Weight : 277.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form White solid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/2-pyridineacetic-acid-5-4-4-5-5-tetramethyl-1-3-2-dioxaborolan-2-yl-methyl-ester.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9806
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart