ADV465748853 AChemBlock

4,6-Dimethyl-2,3-dihydropyridazin-3-one

  • MDL number MFCD00867544
  • Cas Number : 7007-92-3
  • Molecular Weight : 124.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/4-6-dimethyl-2-3-dihydropyridazin-3-one.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9807
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart