ADV465748866 AChemBlock

5-Chloro-3-methyl-1H-pyrazolo[4,3-b]pyridine

  • MDL number MFCD14706407
  • Cas Number : 864775-64-4
  • Molecular Weight : 167.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/5-chloro-3-methyl-1h-pyrazolo-4-3-b-pyridine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9832
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart