ADV465748869 AChemBlock

Methyl 2,2-dimethyl-3-hydroxycyclobutanecarboxylate

  • MDL number MFCD23106161
  • Cas Number : 527751-16-2
  • Molecular Weight : 158.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/methyl-2-2-dimethyl-3-hydroxycyclobutanecarboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9838
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart