ADV465748872 AChemBlock

(R)-1-Methyl-3-(aminomethyl)pyrrolidine

  • MDL number MFCD23106375
  • Cas Number : 1207541-15-8
  • Molecular Weight : 114.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/r-1-methyl-3-aminomethyl-pyrrolidine.html

Safety Information

signalword Danger
pcodes P210;P233;P301+330+331;P303+361+351
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9844
pictograms GHS02; GHS05; GHS09
hstatements H226;H302;H312;H314;H318
Availability details for
Quantity
  • Availability
  • Add to Cart